Merck
  • The first asymmetric halogen/metal-exchange reaction: desymmetrization of alcohols with enantiotopic bromoarene substituents.
The first asymmetric halogen/metal-exchange reaction: desymmetrization of alcohols with enantiotopic bromoarene substituents.
Chemistry (Weinheim an der Bergstrasse, Germany) (2009-06-06)
Daniel Sälinger, Reinhard Brückner
ABSTRACT
Desymmetrizations of the prochiral bis(bromoaryl)alcohols 1 and 4 were effected by treatment with iPr2Mg and enantiomerically pure lithium alkoxides. The resulting arylmagnesium compounds were quenched with various electrophiles. The absolute and (if relevant) relative configurations of the resulting products were determined. The best ee/yield combination was obtained for the protonolysis furnishing monobromoalcohol (R)-2 (53 % ee, 51 % yield). The latter was converted into (R)-orphenadrine, an antihistaminic and anticholinergic drug.
Sigma-Aldrich
Orphenadrine hydrochloride, ≥98.0% (AT)
Orphenadrine for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Orphenadrine citrate salt, analytical standard