Merck
  • Discovery process and pharmacological characterization of 2-(S)-(4-fluoro-2-methylphenyl)piperazine-1-carboxylic acid [1-(R)-(3,5-bis-trifluoromethylphenyl)ethyl]methylamide (vestipitant) as a potent, selective, and orally active NK1 receptor antagonist.
Discovery process and pharmacological characterization of 2-(S)-(4-fluoro-2-methylphenyl)piperazine-1-carboxylic acid [1-(R)-(3,5-bis-trifluoromethylphenyl)ethyl]methylamide (vestipitant) as a potent, selective, and orally active NK1 receptor antagonist.
Journal of medicinal chemistry (2009-04-25)
Romano Di Fabio, Cristiana Griffante, Giuseppe Alvaro, Giorgio Pentassuglia, Domenica A Pizzi, Daniele Donati, Tino Rossi, Giuseppe Guercio, Mario Mattioli, Zadeo Cimarosti, Carla Marchioro, Stefano Provera, Laura Zonzini, Dino Montanari, Sergio Melotto, Philip A Gerrard, David G Trist, Emiliangelo Ratti, Mauro Corsi
ABSTRACT
In an effort to discover novel druglike NK(1) receptor antagonists a new series of suitably substituted C-phenylpiperazine derivatives was identified by an appropriate chemical exploration of related N-phenylpiperazine analogues, with the specific aim to maximize their in vitro affinity and optimize in parallel their pharmacokinetic profile. Among the compounds synthesized, 2-(S)-(4-fluoro-2-methylphenyl)piperazine-1-carboxylic acid [1-(R)-(3,5-bis-trifluoromethylphenyl)ethyl]methylamide (vestipitant) was identified as one of the most in vitro potent and selective NK(1) receptor antagonists ever discovered, showing appropriate pharmacokinetic properties and in vivo activity. On the basis of its preclinical profile, this compound was selected as a drug candidate.