Merck
Mild and efficient DBU-catalyzed amidation of cyanoacetates.
Organic letters (2009-04-04)
Kristin E Price, Claude Larrivée-Aboussafy, Brett M Lillie, Robert W McLaughlin, Jason Mustakis, Kevin W Hettenbach, Joel M Hawkins, Rajappa Vaidyanathan
ABSTRACT
A mild, high-yielding, and practical protocol for the direct amidation of alkyl cyanoacetates using DBU is presented. This method eliminates the need for activation of cyanoacetic acid and/or high temperatures. It has been applied to the large-scale synthesis of CP-690,550-10 (1), a compound under development for the treatment of autoimmune diseases.
Sigma-Aldrich
1,8-Diazabicyclo[5.4.0]undec-7-ene, puriss., ≥99.0% (GC)
Sigma-Aldrich
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98%
Sigma-Aldrich
Cyanoacetic acid, 99%