Merck
Fluoride-triggered indium-mediated synthesis of (hetero)biaryls.
Dalton transactions (Cambridge, England : 2003) (2009-03-26)
Enrique Font-Sanchis, Angela Sastre-Santos, Fernando Fernández-Lázaro
ABSTRACT
A simple method for the preparation of triorganoindium reagents under mild conditions based in indium-silicon exchange is described.
Sigma-Aldrich
Indium(III) chloride, anhydrous, powder, ≥99.999% trace metals basis
Sigma-Aldrich
Indium(III) chloride, 99.999% trace metals basis
Sigma-Aldrich
Indium(III) chloride, 98%