Merck
  • Genetic alterations in K-ras and p53 cancer genes in lung neoplasms from B6C3F1 mice exposed to cumene.
Genetic alterations in K-ras and p53 cancer genes in lung neoplasms from B6C3F1 mice exposed to cumene.
Toxicologic pathology (2008-07-24)
Hue-Hua L Hong, Thai-Vu T Ton, Yongbaek Kim, Nobuko Wakamatsu, Natasha P Clayton, Po-Chuen Chan, Robert C Sills, Stephanie A Lahousse
ABSTRACT
The incidences of alveolar/bronchiolar adenomas and carcinomas in cumene-treated B6C3F1 mice were significantly greater than those of the control animals. We evaluated these lung neoplasms for point mutations in the K-ras and p53 genes that are often mutated in humans. K-ras and p53 mutations were detected by cycle sequencing of PCR-amplified DNA isolated from paraffin-embedded neoplasms. K-ras mutations were detected in 87% of cumene-induced lung neoplasms, and the predominant mutations were exon 1 codon 12 G to T transversions and exon 2 codon 61 A to G transitions. P53 protein expression was detected by immunohistochemistry in 56% of cumene-induced neoplasms, and mutations were detected in 52% of neoplasms. The predominant mutations were exon 5, codon 155 G to A transitions, and codon 133 C to T transitions. No p53 mutations and one of seven (14%) K-ras mutations were detected in spontaneous neoplasms. Cumene-induced lung carcinomas showed loss of heterozygosity (LOH) on chromosome 4 near the p16 gene (13%) and on chromosome 6 near the K-ras gene (12%). No LOH was observed in spontaneous carcinomas or normal lung tissues examined. The pattern of mutations identified in the lung tumors suggests that DNA damage and genomic instability may be contributing factors to the mutation profile and development of lung cancer in mice exposed to cumene.
Product Description
Supelco
Cumene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Cumene, 98%
Supelco
Cumene, analytical standard
Supelco
Cumene, Pharmaceutical Secondary Standard; Certified Reference Material