Merck
[Chemical constituents of Changium smyrnioides].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2008-07-01)
Dong-chun Ren, Shi-hui Qian, Nian-yun Yang, Ning Xie, Jin-ao Duan
ABSTRACT
To study chemical constituents of Changium smyrnioides Wolff. The chemical components were isolated and purified by silica gel column and recrystallization. The chemical structures were elucidated on the basis of physico-chemical properties and spectral data. Ten compounds were isolated and identified as lignoceric acid (1), beta-sitosterol (2), stigmasterol (3), 5-hydroxy-8-methoxypsoralen (4), glycerylmonopalmitate (5), L-pyroglutamic acid (6), succinic acid (7), vanillic acid-4-O-beta-D-glucopyranoside (8 ), vanillic acid (9), daucosterol (10). Compounds 1, 4, 5, 6, 8 and 9 are obtained from the plant for the first time.
Sigma-Aldrich
Lignoceric acid, ≥99% (GC)