Merck
  • Quantitative ester analysis in cachaca and distilled spirits by gas chromatography-mass spectrometry (GC-MS).
Quantitative ester analysis in cachaca and distilled spirits by gas chromatography-mass spectrometry (GC-MS).
Journal of agricultural and food chemistry (2008-06-24)
Eduardo S P Nascimento, Daniel R Cardoso, Douglas W Franco
ABSTRACT
An analytical procedure for the separation and quantification of ethyl acetate, ethyl butyrate, ethyl hexanoate, ethyl lactate, ethyl octanoate, ethyl nonanoate, ethyl decanoate, isoamyl octanoate, and ethyl laurate in cachaca, rum, and whisky by direct injection gas chromatography-mass spectrometry was developed. The analytical method is simple, selective, and appropriated for the determination of esters in distilled spirits. The limit of detection ranged from 29 (ethyl hexanoate) to 530 (ethyl acetate) microg L(-1), whereas the standard deviation for repeatability was between 0.774% (ethyl hexanoate) and 5.05% (isoamyl octanoate). Relative standard deviation values for accuracy vary from 90.3 to 98.5% for ethyl butyrate and ethyl acetate, respectively. Ethyl acetate was shown to be the major ester in cachaca (median content of 22.6 mg 100 mL(-1) anhydrous alcohol), followed by ethyl lactate (median content of 8.32 mg 100 mL(-1) anhydrous alcohol). Cachaca produced in copper and hybrid alembic present a higher content of ethyl acetate and ethyl lactate than those produced in a stainless-steel column, whereas cachaca produced by distillation in a stainless-steel column present a higher content of ethyl octanoate, ethyl decanoate, and ethyl laurate. As expected, ethyl acetate is the major ester in whiskey and rum, followed by ethyl lactate for samples of rum. Nevertheless, whiskey samples exhibit ethyl lactate at contents lower or at the same order of magnitude of the fatty esters.
Sigma-Aldrich
(−)-Ethyl L-lactate, 98%
Sigma-Aldrich
Ethyl lactate, ≥98%, FCC, FG
Sigma-Aldrich
(−)-Ethyl L-lactate, purum, ≥98.0% (sum of enantiomers, GC)
Sigma-Aldrich
Ethyl lactate, natural, ≥98%, FCC, FG
Sigma-Aldrich
(−)-Ethyl L-lactate, ≥99.0%
Sigma-Aldrich
Ethyl nonanoate, 97%