Merck
  • MRP7/ABCC10 expression is a predictive biomarker for the resistance to paclitaxel in non-small cell lung cancer.
MRP7/ABCC10 expression is a predictive biomarker for the resistance to paclitaxel in non-small cell lung cancer.
Molecular cancer therapeutics (2008-05-01)
Tetsuya Oguri, Hiroaki Ozasa, Takehiro Uemura, Yuji Bessho, Mikinori Miyazaki, Ken Maeno, Hiroyoshi Maeda, Shigeki Sato, Ryuzo Ueda
ABSTRACT
We used the paclitaxel-resistant human small cell lung cancer subline PC-6/TAX1-1, selected from PC-6 cells by paclitaxel, to test whether MRP7/ABCC10 (ABCC10) confers paclitaxel resistance. We found that gene expression of both ABCB1/MDR1 (ABCB1) and ABCC10 was higher in PC-6/TAX1-1 cells than in PC-6 cells. The expression levels of ABCC10 showed a significant inverse correlation with paclitaxel sensitivity (r = 0.574; P < 0.05) in 17 non-small cell lung cancer (NSCLC) cells unlike the expression levels of ABCB1. Pretreatment with the ABCC10 inhibitor sulfinpyrazone altered the sensitivity to paclitaxel in ABCC10-expressing NSCLC cells, concomitant with increased intracellular paclitaxel accumulation. These findings suggest that expression of the ABCC10 gene is induced by paclitaxel and that ABCC10 confers paclitaxel resistance by enhancing the efflux for paclitaxel. To confirm this hypothesis, we tested the effect on paclitaxel cytotoxicity of decreasing the expression of ABCC10 by small interfering RNA and found that this enhanced paclitaxel cytotoxicity in NCI-H23 cells concomitant with increased intracellular paclitaxel accumulation. These data indicate that ABCC10 may be one of the biomarkers for paclitaxel resistance in NSCLC.
Product Description
Supelco
(±)-Sulfinpyrazone, analytical standard