Merck
  • Vibrational spectra of small silicon monoxide cluster cations measured by infrared multiple photon dissociation spectroscopy.
Vibrational spectra of small silicon monoxide cluster cations measured by infrared multiple photon dissociation spectroscopy.
Physical chemistry chemical physics : PCCP (2008-03-11)
Etienne Garand, Daniel Goebbert, Gabriele Santambrogio, Ewald Janssens, Peter Lievens, Gerard Meijer, Daniel M Neumark, Knut R Asmis
ABSTRACT
The first gas-phase infrared spectra of silicon monoxide cations (SiO)(n)(+), n = 3-5, using multiple photon dissociation in the 550-1250 cm(-1) frequency range, are reported. All clusters studied here fragment via loss of a neutral SiO unit. The experimental spectra are compared to simulated linear absorption spectra from calculated low energy isomers for each cluster. This analysis indicates that a "ring" isomer is the primary contributor to the (SiO)(3)(+) spectrum, that the (SiO)(4)(+) spectrum results from two close-lying bicyclic ring isomers, and that the (SiO)(5)(+) spectrum is from a bicyclic ring with a central, fourfold-coordinated Si atom. Experiment and theory indicate that the energies and energetic orderings of (SiO)(n)(+) isomers differ from those for neutral (SiO)(n) clusters.
Sigma-Aldrich
Silicon monoxide, pieces, 3-10 mm, 99.99% trace metals basis
Sigma-Aldrich
Silicon monoxide, powder, −325 mesh