Merck
  • An enantioselective biginelli reaction catalyzed by a simple chiral secondary amine and achiral brønsted acid by a dual-activation route.
An enantioselective biginelli reaction catalyzed by a simple chiral secondary amine and achiral brønsted acid by a dual-activation route.
Chemistry (Weinheim an der Bergstrasse, Germany) (2008-02-05)
Junguo Xin, Lu Chang, Zongrui Hou, Deju Shang, Xiaohua Liu, Xiaoming Feng
ABSTRACT
An enantioselective Biginelli reaction that proceeds by a dual-activation route has been developed by using a combined catalyst of a readily available trans-4-hydroxyproline-derived secondary amine and a Brønsted acid. Aromatic, heteroaromatic, and fused-ring aldehydes were found to be suitable substrates for this multicomponent reaction. The corresponding dihydropyrimidines were obtained in moderate-to-good yields with up to 98 % ee under mild conditions. Based on the experimental results and the observed absolute configurations of the products, a plausible transition state has been proposed to explain the origin of the activation and the asymmetric induction.
Sigma-Aldrich
Ethyl acetoacetate, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Ethyl acetoacetate, Arxada quality, ≥99.0% (GC)
Sigma-Aldrich
Ethyl acetoacetate, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl acetoacetate, natural, ≥97%, FG
Sigma-Aldrich
Ethyl acetoacetate, ≥99%, FCC, FG