Merck
Fragrance material review on cis-beta-damascone.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-27)
J Lalko, A Lapczynski, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of cis-beta-damascone when used as a fragrance ingredient is presented.