Merck
Fragrance material review on phenyl salicylate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-23)
A Lapczynski, L Jones, D McGinty, S P Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of phenyl salicylate when used as a fragrance ingredient is presented.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 30034252, Phenyl salicylate, traceable to primary standards
Supelco
Phenyl salicylate melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenyl salicylate, ≥99%, FG
Sigma-Aldrich
Phenyl salicylate, ReagentPlus®, 99%