MilliporeSigma
  • Synthesis of some new 4,5-substituted-4H-1,2,4-triazole-3-thiol derivatives.
Synthesis of some new 4,5-substituted-4H-1,2,4-triazole-3-thiol derivatives.
Molecules (Basel, Switzerland) (2007-11-17)
A Cansiz, M Koparir, A Demirdağ
ABSTRACT
In this study appropriate hydrazide compounds, furan-2-carboxylic acid hydrazide (1) and phenylacetic acid hydrazide (2) were converted into 1,4-substituted thiosemicarbazides 4a-e and 5a-e and 4-amino-5-(furan-2-yl or benzyl)-4H-1,2,4-triazole-3-thiols 7 and 10. The 1,4-substituted thiosemicarbazides were then converted into 5-(furan-2-yl or benzyl)-4-(aryl)-4H-1,2,4-triazole-3-thiols 8a-e and 9a-e. In addition, the azomethines 11a-d and 12a-d were prepared from the corresponding arylaldehydes and the 4-amino-5-(furan-2-yl or benzyl)-4H-1,2,4-triazole-3-thiols 7 and 10. The structures of all the synthesized compounds were confirmed by elemental analyses, IR, (1)H-NMR and(13) C-NMR spectra.
Sigma-Aldrich
2-Furoic acid, 98%