Merck
  • Synthesis of novel N-(4-ethoxyphenyl) azetidin-2-ones and their oxidative N-deprotection by ceric ammonium nitrate.
Synthesis of novel N-(4-ethoxyphenyl) azetidin-2-ones and their oxidative N-deprotection by ceric ammonium nitrate.
Molecules (Basel, Switzerland) (2007-11-06)
Aliasghar Jarrahpour, Maaroof Zarei
ABSTRACT
It is shown that the N-(p-ethoxyphenyl) group on beta-lactams can be oxidatively removed by ceric ammonium nitrate in good yield. Fourteen new N-(p-ethoxyphenyl)-2-azetidinones 8a-n were synthesized through standard [2+2] ketene-imine cycloadditions (Staudinger reaction). Treatment of these compounds with ceric ammonium nitrate yielded the N-dearylated 2-azetidinones 9a-n in good to excellent yields. The effects of solvent, molar equiv of CAN and different temperatures have been investigated and optimum conditions were established.
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ACS reagent, ≥98.5%
Sigma-Aldrich
Ammonium cerium(IV) nitrate, puriss. p.a., ACS reagent, ≥98.5% (RT)
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥98% (titration)