Merck
  • Palladate salts from chiral pyridinium ionic liquids: synthesis and crystal structures.
Palladate salts from chiral pyridinium ionic liquids: synthesis and crystal structures.
Molecules (Basel, Switzerland) (2007-10-26)
Emmanuel Tourneux, Heinz Gornitzka, Jean-Daniel Marty, Nancy Lauth-de Viguerie
ABSTRACT
This paper reports the preparation, characterization and the crystal X-ray structures of metal-containing ionic liquid complexes based on chiral pyridinium cations and tetrachloropalladate (II) [PdCl4]2-.
Sigma-Aldrich
Ammonium tetrachloropalladate(II), ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium tetrachloropalladate(II), 97%
Sigma-Aldrich
Potassium tetrachloropalladate(II), 98%
Sigma-Aldrich
Sodium tetrachloropalladate(II), ≥99.99% trace metals basis
Sigma-Aldrich
Sodium tetrachloropalladate(II), 98%
Sigma-Aldrich
Potassium tetrachloropalladate(II), 99.99% trace metals basis