- Syntheses and binding affinities of 6-nitroquipazine analogues for serotonin transporter. Part 5: 2'-Substituted 6-nitroquipazines.
Syntheses and binding affinities of 6-nitroquipazine analogues for serotonin transporter. Part 5: 2'-Substituted 6-nitroquipazines.
Bioorganic & medicinal chemistry (2007-03-23)
Jae Hak Lee, Yong Hyun Choi, Yoo Jin Lim, Byoung Se Lee, Dae Yoon Chi, Changbae Jin
PMID17376695
ABSTRACT
Five C2'-substituted 6-nitroquipazine (6-NQ) derivatives were prepared and evaluated in terms of their biological abilities (K(i)) to displace [(3)H]citalopram binding to serotonin transporter. The relationship between their structure and biological activities revealed that shorter alkyl groups tend to possess higher binding affinity. Both compounds 12a and 12c were found to have the equally highest binding affinity (K(i)=0.43+/-0.02 nM).