Merck
  • Therapeutic drug monitoring of tacrolimus with the dried blood spot method.
Therapeutic drug monitoring of tacrolimus with the dried blood spot method.
Journal of pharmaceutical and biomedical analysis (2006-12-23)
K Hoogtanders, J van der Heijden, M Christiaans, P Edelbroek, J P van Hooff, L M L Stolk
ABSTRACT
In a preliminary investigation an assay for tacrolimus based on fingerprick sampling and consecutive application as a blood spot on sampling paper has been developed. The dried blood spot was analysed by HPLC-tandem mass spectrometry. The validated range was 1-30 microg/l. Intra- and inter-assay variability for precision and accuracy was <7.5% and 15%, respectively. Tacrolimus concentrations of 24 stable out patients were compared after both blood spot sampling and conventional venous sampling. Method agreement was investigated with the methods of Passing and Bablok and Bland Altman and proved suitable for clinical use. The dried blood spot method for tacrolimus seems promising for patient monitoring.
Sigma-Aldrich
Ascomycin from Streptomyces hygroscopicus var. ascomyceticus
Supelco
Ascomycin solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®