Merck
  • Distribution of PAEs in the middle and lower reaches of the Yellow River, China.
Distribution of PAEs in the middle and lower reaches of the Yellow River, China.
Environmental monitoring and assessment (2006-12-13)
Yujuan Sha, Xinghui Xia, Zhifeng Yang, Gordon H Huang
ABSTRACT
Samples of water, sediment and suspended particulates were collected from 13 sites in the middle and lower reaches of the Yellow River in China. Phthalic acid esters (PAEs) concentrations in different phases of each sample were determined by Gas Chromatogram GC-FID. The results are shown as follows: (1) In the Xiao Langdi-Dongming Bridge section, PAEs concentrations in water phase from the main river ranged from 3.99 x 10(-3) to 45.45 x 10(-3) mg/L, which were similar to those from other rivers in the world. The PAEs levels in the tributaries of the Yellow River were much higher than those of the main river. (2) In the studied branches, the concentration of PAEs in sediment for Luoyang Petrochemical Channel (331.70 mg/Kg) was the highest. The concentrations of PAEs in sediment phase of the main river were 30.52 to 85.16 mg/Kg, which were much higher than those from other rivers in the world. In the main river, the concentration level of PAEs on suspended solid phases reached 94.22 mg/Kg, and it reached 691.23 mg/Kg in the Yiluo River - one tributary of the Yellow River. (3) Whether in the sediment or on the suspended solid phases, there was no significant correlation between the contents of PAEs and TOC or particle size of the solid phase; and the calculated Koc of Di (2-Ethylhexyl) Phthalate (DEHP) in the river were much less than the theoretical value, which inferred that PAEs were not on the equilibrium between water and suspended solid phases/sediment. (4) Among the measured PAEs compounds, the proportions of DEHP and di-n-butyl phthalate (DBP) were much higher than the others. The concentrations of DEHP exceeded the Quality Standard in all the main river and tributary stations except those in the Mengjin and Jiaogong Bridge of the main river. This indicates that more attention should be paid to pollution control and further assessment in understanding risks associated with human health.
Sigma-Aldrich
Di-n-octyl phthalate, ≥98.0% (GC)
Supelco
Di-n-octyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Di-n-octyl phthalate, analytical standard