Merck
Sensitive and rapid electrochemical bioassay of glycosidase activity.
The Analyst (2006-10-10)
Tanin Tangkuaram, Jared Q Gerlach, Yun Xiang, Abdel-Nasser Kawde, Zong Dai, Veer P Bhavanandan, Jeffrey T La Belle, Waret Veerasai, Lokesh Joshi, Joseph Wang
ABSTRACT
Here we present a highly sensitive, rapid and simple electrochemical assay for glycosidases based on treatment of the glycosidase with the appropriate p-nitrophenyl glycoside and anodic detection of released p-nitrophenol. The attractive characteristics of the new bioassay should facilitate advanced glycomic research and routine clinical diagnostics since glycosidases are associated with various diseases.
Sigma-Aldrich
4-Nitrophenyl β-D-mannopyranoside, ≥98%