MilliporeSigma
  • In vivo comparison of various polymeric and low molecular mass inhibitors of intestinal P-glycoprotein.
In vivo comparison of various polymeric and low molecular mass inhibitors of intestinal P-glycoprotein.
Biomaterials (2006-08-22)
Florian Föger, Herbert Hoyer, Krum Kafedjiiski, Michael Thaurer, Andreas Bernkop-Schnürch
ABSTRACT
Several polymers have been reported to modulate drug absorption by inhibition of intestinal P-glycoprotein (P-gp). The aim of the present study was to provide a direct in vivo comparison of delivery systems based on Pluronic P85, Myrj 52 and chitosan-4-thiobutylamidine (Ch-TBA) in vivo in rats, using rhodamine-123 (Rho-123) as representative P-gp substrate. Furthermore, the postulated low molecular mass P-gp inhibitors 6-mercaptopurine and reduced glutathione (GSH) were evaluated in vitro and in vivo. In vitro, the permeation enhancing effect of 6-mercaptopurine, GSH, Pluronic P85, Myrj 52, and the combination of Ch-TBA with GSH was evaluated by using freshly excised rat intestinal mucosa mounted in Ussing-type diffusion chambers. In comparison to buffer only, Rho-123 transport in presence of 100 microm 6-mercaptopurine, 0.5% (w/v) GSH, 0.5% (w/v) Pluronic P85, 0.5% (w/v) Myrj 52 and the combination of 0.5% (w/v) Ch-TBA/ 0.5% (w/v) GSH, was 2.1, 1.6, 1.9, 1.8, 3.0-fold improved, respectively. In vivo in rat, enteric-coated tablets based on Pluronic P85, Myrj 52 or Ch-TBA/GSH increased the area under the plasma concentration time curve (AUC(0-12)) of Rho-123 1.6-fold, 2.4-fold, 4.3-fold, respectively, in comparison to control only. Contrariwise, the low molecular mass excipients 6-mercaptopurine and GSH showed no significant effect in vivo at all. This in vivo study showed that polymeric P-gp inhibitors and especially the delivery system based on thiolated chitosan significantly increased the oral bioavailability of P-gp substrate Rho-123.
Sigma-Aldrich
Polyoxyethylene (40) stearate