Merck
  • Characterization of recombinant human cardiac KCNQ1/KCNE1 channels (I (Ks)) stably expressed in HEK 293 cells.
Characterization of recombinant human cardiac KCNQ1/KCNE1 channels (I (Ks)) stably expressed in HEK 293 cells.
The Journal of membrane biology (2006-08-16)
Ming-Qing Dong, Chu-Pak Lau, Zhan Gao, Gea-Ny Tseng, Gui-Rong Li
ABSTRACT
The present study was designed to characterize pharmacological, biophysical and electrophysiological properties of the recombinant human cardiac I (Ks) (KCNQ1/KCNE1) channels at physiological temperature. Human cardiac KCNQ1 and KCNE1 genes were cotransfected into HEK 293 cells, and a cell clone stably expressing both genes was selected. Membrane currents were recorded using a perforated patch-clamp technique. The typical I (Ks) was slowly activated upon depolarization voltages in HEK 293 cells stably expressing human cardiac KCNQ1 and KCNE1 genes, and the current was inhibited by I (Ks) blockers HMR 1556 and chromanol 293B, with 50% inhibitory concentrations (IC(50)s) of 83.8 nM: and 9.2 muM: , respectively. I (Ks) showed a significant temperature-dependent increase in its magnitude upon elevating bath temperature to 36 degrees C from room temperature (21 degrees C). The current was upregulated by the beta-adrenoceptor agonist isoproterenol, and the effect was reversed by H89. In addition, I (Ks) was inhibited by Ba(2+) in a concentration-dependent manner (IC(50) = 1.4 mM). Action potential clamp revealed a "bell-shaped" time course of I (Ks) during the action potential, and maximal peak current was seen at the plateau of the action potential. A significant use- and frequency-dependent increase of I (Ks) was observed during a train of action potential clamp. These results indicate that the recombinant human cardiac I (Ks) stably expressed in HEK 293 cells is similar to native I (Ks) in drug sensitivity and regulated by Ba(2+) and beta-adrenoceptor via the cyclic adenosine monophosphate/protein kinase A pathway. Importantly, the current exhibits significant temperature dependence, a bell-shaped time course during action potential and prominent use- or frequency-dependent accumulation during a train of action potentials.
Sigma-Aldrich
Chromanol 293B, ≥98% (HPLC), powder