Merck
  • Severe anaphylactic reaction after intracameral antibiotic administration during cataract surgery.
Severe anaphylactic reaction after intracameral antibiotic administration during cataract surgery.
Journal of cataract and refractive surgery (2005-04-07)
José R Villada, Ubaldo Vicente, Jaime Javaloy, Jorge L Alió
ABSTRACT
We report a severe anaphylactic reaction that occurred about 5 minutes after 1.0 mg of cefuroxime was injected into the anterior chamber after routine phacoemulsification and intraocular lens implantation. The patient was known to be allergic to penicillin. Immediate action was taken, and the patient recovered well. The presence of staff trained in resuscitation was essential in this case and raises questions about the trend to perform routine topical anesthesia cataract surgery without an anesthesiologist in the operating room.
Product Description
Supelco
Chlorpheniramine Maleate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
S-(+)-Chlorpheniramine maleate salt
Dexchlorpheniramine maleate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-Chlorpheniramine maleate salt, ≥99% (perchloric acid titration)
Chlorphenamine maleate, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorpheniramine maleate salt solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®