Merck
  • [Chiral separation of 3-phenyllactic acid by capillary electrophoresis].
[Chiral separation of 3-phenyllactic acid by capillary electrophoresis].
Se pu = Chinese journal of chromatography (2005-02-17)
Demao Li, Congfa Li, Sixin Liu, Limei Chen
ABSTRACT
Various factors affecting the chiral resolution of 3-phenyllactic acid by capillary zone electrophoresis such as the type of cyclodextrin, concentration of the chiral selector, buffer pH, voltage, and temperature were studied and optimized. Using 30 mmol/L hydroxypropyl-beta-cyclodextrin as the chiral selector, and 0.1 mol/L phosphate solution (pH 5.5) as the running buffer, the baseline separation of 3-phenyllactic acid enantiomers was achieved with a resolution factor of 1.51. The separation voltage was 26 kV and the temperature was 25 degrees C.
Sigma-Aldrich
L-(−)-3-Phenyllactic acid, 98%
Sigma-Aldrich
D-(+)-3-Phenyllactic acid, 98%
Sigma-Aldrich
3-Phenyllactic acid, ≥98%