Merck
[Studies on chemical constituents of Pholidota yunnanensis].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2005-02-16)
Zhi-ming Bi, Zheng-tao Wang, Luo-shan Xu, Guo-jun Xu
ABSTRACT
To investigate the chemical constituents from Pholidota yunnanensis. Various chromatographic techniques were employed for isolation and purification of the constituents. The structures were elucidated by chemical and spectral analyses. Seven compounds were obtained and they were identified by spectroscopic analysis as n-nonacosane, cyclopholidone, n-dotriacontanoic acid, n-octacostyl ferulate, cyclopholidonol, cycloneolitsol and beta-sitosterol, respectively. n-octacostyl ferulate and cycloneolitsol were isolated from genus Pholidota for the first time.