Merck
  • Double N-arylation of primary amines: carbazole synthesis from 2,2'-biphenyldiols.
Double N-arylation of primary amines: carbazole synthesis from 2,2'-biphenyldiols.
The Journal of organic chemistry (2005-01-18)
Atsushi Kuwahara, Koji Nakano, Kyoko Nozaki
ABSTRACT
The double N-arylation of primary amines with 2,2'-biphenylylene ditriflates was investigated for the synthesis of multisubstituted carbazoles. Palladium complexes supported by 2-dicyclohexylphosphino-2'-methylbiphenyl or Xantphos [4,5-bis(diphenylphosphino)-9,9-dimethylxanthene] were found to be efficient catalysts for the reaction. The catalysts allow the use of anilines with an electron-donating or electron-withdrawing substituent and multisubstituted 2,2'-biphenylylene ditriflates as substrates. Ammonia equivalents, such as O-tert-butyl carbamate, are also employable as a nitrogen source to give the N-protected carbazoles which can easily give the corresponding N-unsubstituted carbazoles after deprotection. By using this methodology, a carbazole alkaloid, mukonine, is synthesized in 40% yield for five steps, in comparable efficiency to the recent precedents.
Sigma-Aldrich
2,2′-Biphenol, 99%