Merck
Kinetics and thermodynamics of hexameric capsule formation.
Journal of the American Chemical Society (2004-03-05)
Masamichi Yamanaka, Alexander Shivanyuk, Julius Rebek
ABSTRACT
Resorcinarene 1b forms a hexameric assembly in water-saturated CDCl(3) that encapsulates one tetraalkylammonium salt (2(+)Br(-)). The remaining space is occupied by coencapsulated solvent molecules. A maximum of three and minimum of one CHCl(3) molecule were found inside of capsules with tetrapropyl- and tetraheptylammonium bromide, respectively. The encapsulation of the salt is endothermic and entropically favored by the liberation of the solvent molecules. The stabilities of the encapsulation complexes and the rates of guest exchange decrease for larger cations. The higher activation barriers for in/out exchange of the larger guests suggest tight conformational restraints in the transition state. It is likely that complete dissociation of one resorcinarene molecule is necessary for the exchange of the alkylammonium salts.
Sigma-Aldrich
Tetrahexylammonium chloride, 96%
Sigma-Aldrich
Tetrahexylammonium bromide, 99%
Sigma-Aldrich
Tetrahexylammonium hydroxide solution, ~40% in H2O (T)
Supelco
Tetrahexylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetrahexylammonium hydrogensulfate, ≥98.0% (T)
Supelco
Tetrahexylammonium hydrogensulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)