Merck
Secondary metabolites from the liverwort Tylimanthus renifolius.
Phytochemistry (2003-11-25)
Hildegard Feld, Josef Zapp, Hans Becker
ABSTRACT
Two new herbertane sesquiterpenoids, (-)-herbertene-1,12-diol and (-)-gamma-herbertenol, a new aromadendrane, 9-acetoxy-10-hydroxyaromadendrane, a new rosane diterpenoid, 5,15-rosadiene-3,11-dione and a new phytane, 2Z-phytene-1,15,20-triol, along with the known (-)-alpha-herbertenol have been isolated from the liverwort Tylimanthus renifolius. Several flavones and flavanones with an unsubstituted ring B were also identified. This is the first report of this class of flavonoids from the Hepaticae. All structures were elucidated by NMR spectroscopy.