Merck
  • Remineralization effect of a low-concentration fluoride rinse in an intraoral model.
Remineralization effect of a low-concentration fluoride rinse in an intraoral model.
Caries research (2002-05-31)
L C Chow, S Takagi, S Frukhtbeyn, B A Sieck, E E Parry, N S Liao, G E Schumacher, M Markovic
ABSTRACT
A previous study showed that a sodium hexafluorosilicate-calcium chloride-based two-solution fluoride (F) rinse containing 6 mmol/l of F was more effective than a 12 mmol/l F sodium fluoride rinse in depositing F on tooth surfaces and increasing oral F levels. The present study compared the remineralization effects of these two rinses in an intraoral de- and remineralization model. The results showed that the 6 mmol/l F two-solution rinse produced greater remineralization in increasing lesion mineral contents and reducing lesion depths. The results demonstrated that the effectiveness of an F regimen depends less on the F dose and more on the ability of the treatment to utilize F efficiently for remineralization.
Product Description
Supelco
Potassium hexafluorosilicate, ≥99.0% (T)