Merck
  • New possible chelating agents of clinical interest for iron(III): 2-hydroxynicotinic acid and 3-hydroxypicolinic acid. A thermodynamic study.
New possible chelating agents of clinical interest for iron(III): 2-hydroxynicotinic acid and 3-hydroxypicolinic acid. A thermodynamic study.
Annali di chimica (2002-01-05)
V B Di Marco, A Tapparo, G G Bombi
ABSTRACT
The formation of complexes between iron(III) and two ligands of possible relevance to chelation therapy, 2-hydroxynicotinic acid and 3-hydroxypicolinic acid, in aqueous 0.6 m (Na)Cl at 25 degrees C, has been investigated by means of potentiometric titrations and UV measurements. In both cases several mononuclear complexes and their deprotonation products are formed.
Sigma-Aldrich
3-Hydroxypyridine-2-carboxylic acid, 98%
Supelco
3-Hydroxypicolinic acid, ≥99.0% (HPLC)
Sigma-Aldrich
2-Hydroxypyridine-3-carboxylic acid, 98%