Merck
  • Synergistic insulinotropic effects of succinic acid dimethyl ester and exendin-4 in anaesthetized rats.
Synergistic insulinotropic effects of succinic acid dimethyl ester and exendin-4 in anaesthetized rats.
International journal of molecular medicine (2001-08-09)
J Cancelas, M L Villanueva-Peñacarrillo, I Valverde, W J Malaisse
ABSTRACT
It was recently proposed that suitable succinic acid esters could be used to potentiate the insulinotropic action of glucagon-like peptide 1 (GLP-1) in the treatment of type-2 diabetes mellitus. In such a perspective, the present study aimed mainly at investigating whether exendin-4 (Ex-4), a peptide structurally related to GLP-1(7-36)amide, and succinic acid dimethyl ester (SAD) also act synergistically upon insulin secretion in anaesthetized rats. Despite a higher plasma insulin concentration in SAD-infused rats (5.5+/-1.1 ng/ml) than in saline-infused animals (1.9+/-0.7 ng/ml), the intravenous injection of Ex-4 augmented to a greater extent the plasma concentration of insulin in the former rats (+7.4+/-2.5 ng/ml) than in the latter animals (+2.8+/-0.6 ng/ml). These findings document that the insulinotropic actions of Ex-4 and GLP-1 display comparable nutrient dependency, being both potentiated by a non-glucidic nutrient secretagogue such as SAD.
Sigma-Aldrich
DBE-4 dibasic ester, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl succinate, 98%, FG