Merck
  • Reactive oxygen species in the aerobic decomposition of sodium hydroxymethanesulfinate.
Reactive oxygen species in the aerobic decomposition of sodium hydroxymethanesulfinate.
Archives of biochemistry and biophysics (1999-07-09)
S V Makarov, C Mundoma, S A Svarovsky, X Shi, P M Gannett, R H Simoyi
ABSTRACT
Sodium hydroxymethanesulfinate, (HOCH2SO2Na, HMS) is relatively stable in aqueous alkaline environments, but rapidly decomposes in acidic medium to give a variety of products that include sulfur dioxide. A detailed kinetic and mechanistic study of the decomposition of HMS in slightly acidic medium has shown a process that produces dithionite, S2O2-4, which is preceded by an induction period which persists for as long as molecular oxygen is present in the reaction solution. The complete consumption of molecular oxygen is a prerequisite for the formation of S2O2-4. Among some of the intermediates detected in the decomposition of HMS is the sulfite radical, SO-3. Comparisons are made between the decomposition mechanisms of thiourea dioxide (aminoiminomethanesulfinic acid) and HMS.
Sigma-Aldrich
Hydroxymethanesulfinic acid monosodium salt dihydrate
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxymethanesulfinate dihydrate, ≥98.0% (RT)
Pricing and availability is not currently available.