Merck
  • Beta-endorphin and the immune system--possible role in autoimmune diseases.
Beta-endorphin and the immune system--possible role in autoimmune diseases.
Autoimmunity (1995-01-01)
H Mørch, B K Pedersen
ABSTRACT
The immune system and the neuroendocrine system are closely interconnected having such means of bidirectional communication and regulation. In this review, a hypothesis is put forward regarding the possible role of beta-endorphins in the pathogenesis of autoimmune diseases: It is suggested that the increased cytokine production in immunoinflammatory disorders induces production of beta-endorphins from the pituitary and the lymphocytes; the enhanced level of beta-endorphin causes inhibition of human T helper cell function, which potentially down-regulate the antibody production. Also the beta-endorphin-induced enhancement of the natural killer cell activity may suppress the B cell function. In addition, beta-endorphin also exerts a direct inhibitory effect on the antibody production. Thus, in autoimmune disorders the enhanced cytokine level may via stimulation of the production of beta-endorphins exert a negative feed back on the antibody production and potentially so on the production of autoantibodies.
Sigma-Aldrich
β-Endorphin human, ≥95% (HPLC)