Merck
  • Antihyperlipidaemic activity of swertiamarin, a secoiridoid glycoside in poloxamer-407-induced hyperlipidaemic rats.
Antihyperlipidaemic activity of swertiamarin, a secoiridoid glycoside in poloxamer-407-induced hyperlipidaemic rats.
Journal of natural medicines (2009-07-28)
Hitesh Vaidya, Mandapati Rajani, Vasudevan Sudarsanam, Harish Padh, Ramesh Goyal
ABSTRACT
We have investigated antihyperlipidaemic effect of swertiamarin (50 mg/kg, oral once) isolated from the perennial herb Enicostemma littorale Blume in poloxamer 407 (P-407)-induced hyperlipidaemic rats. Rats were made hyperlipidaemic by intraperitoneal administration of P-407 (400 mg/kg). Serum lipid levels such as total cholesterol, triglycerides and low-density lipoprotein cholesterol increased significantly (P < 0.001) compared with normal control rats. All these changes were significantly prevented in the rats treated with swertiamarin. Serum high-density lipoprotein (HDL) cholesterol was found to be reduced in the P-407 control rats. However, administration of swertiamarin significantly (P < 0.01) increased HDL levels and it showed a significant lipid-lowering effect, as well as a high antiatherogenic potential. Overall swertiamarin is an effective lipid-lowering lead compound and can be useful for preventing atherosclerosis.
Sigma-Aldrich
Swertiamarin, ≥95% (LC/MS-ELSD)
Supelco
Swertiamarin, analytical standard