Merck
  • Molecular cloning and chromosomal localization of the ADH7 gene encoding human class IV (sigma) ADH.
Molecular cloning and chromosomal localization of the ADH7 gene encoding human class IV (sigma) ADH.
Genomics (1996-01-15)
H Yokoyama, E Baraona, C S Lieber
ABSTRACT
The ADH7 gene encoding human Class IV (sigma) alcohol dehydrogenase (ADH) was cloned from a Caucasian genomic DNA library and characterized. It has nine exons and eight introns that span about 22 kb, and its intron insertion is identical to that of the other ADH genes (ADH1 to ADH5). The nucleotide sequences of the exons encoding 374 amino acids are identical to the previously reported cDNA sequence of sigma ADH. Fluorescence in situ hybridization analysis showed that ADH7 is located on human chromosome 4q23-q24, close to the ADH cluster locus (4q21-q25). These data are consistent with the view that Class IV ADH is a member of the ADH family and is phylogenetically close to the other ADHs.
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, powder, ≥300 units/mg protein, mol wt ~141,000 (four subunits)
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa
Sigma-Aldrich
Alcohol Dehydrogenase equine, recombinant, expressed in E. coli, ≥0.5 U/mg