- Molecular cloning and nucleotide sequence of human pancreatic prechymotrypsinogen cDNA.
Molecular cloning and nucleotide sequence of human pancreatic prechymotrypsinogen cDNA.
Biochemical and biophysical research communications (1989-01-31)
N Tomita, Y Izumoto, A Horii, S Doi, H Yokouchi, M Ogawa, T Mori, K Matsubara
PMID2917002
ABSTRACT
The cDNA clone encoding human prechymotrypsinogen was isolated from a human pancreas cDNA library and its nucleotide sequence was determined. The sequence consists of a 16 bp 5' non-coding region, a 789 bp amino acid coding region and a 60 bp 3' non-coding region. The predicted product consists of 263 amino acids, including 18 amino acids for a signal peptide and 15 amino acids possible for an activation peptide. Southern blot analyses using the cloned cDNA as a probe revealed that human genomic DNA carries at least two genes that are related to chymotrypsinogen.
Sigma-Aldrich
α-Chymotrypsin−Agarose from bovine pancreas, lyophilized powder, 2,000-3,500 units/g agarose (One ml gel will yield 65-120 units)
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type I-S, essentially salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, suitable for protein sequencing, salt-free, lyophilized powder