Merck
  • Regulators of cellular levels of histone acetylation in Saccharomyces cerevisiae.
Regulators of cellular levels of histone acetylation in Saccharomyces cerevisiae.
Genetics (2008-05-22)
Weimin Peng, Cynthia Togawa, Kangling Zhang, Siavash K Kurdistani
ABSTRACT
Histone acetylation levels are regulated through the opposing activities of histone acetyltransferases (HATs) and deacetylases (HDACs). While much is known about gene-specific control of histone acetylation, little is understood about how total or cellular levels of histone acetylation are regulated. To identify regulators of cellular levels of histone acetylation, we developed an immunofluorescence-based approach to screen the single-gene deletion library of Saccharomyces cerevisiae for strains with significant reductions in cellular histone acetylation levels. Of the 4848 mutants screened, we identified 63 strains with considerable cellular hypoacetylation of N-terminal lysines in histones H3 and H4. The cellular hypoacetylation was validated for subsets of the identified strains through secondary screens including mass spectrometric analysis of individual lysines and chromatin immunoprecipitation of specific genomic loci. Among the identified mutants were several members of the Ccr4-Not complex, V-type ATPases, and vacuolar protein-sorting complexes as well as genes with unknown functions. We show that Gcn5, a major HAT in yeast, has diminished histone acetyltransferase activity in particular mutants, providing a plausible explanation for reduction of cellular acetylation levels in vivo. Our findings have revealed unexpected and novel links between histone acetylation, Gcn5 HAT activity, and diverse processes such as transcription, cellular ion homeostasis, and protein transport.
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Tanshinone IIA, ≥97% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Poly-L-lysine solution, 0.1 % (w/v) in H2O
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, ≥99.0% (GC)
Supelco
Total Phenolics, certified reference material
Sigma-Aldrich
Poly-L-lysine solution, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly-L-lysine solution, mol wt 150,000-300,000, 0.01%, sterile-filtered, BioReagent, suitable for cell culture