Merck
The DNA sequence, annotation and analysis of human chromosome 3.
Nature (2006-04-28)
Donna M Muzny, Steven E Scherer, Rajinder Kaul, Jing Wang, Jun Yu, Ralf Sudbrak, Christian J Buhay, Rui Chen, Andrew Cree, Yan Ding, Shannon Dugan-Rocha, Rachel Gill, Preethi Gunaratne, R Alan Harris, Alicia C Hawes, Judith Hernandez, Anne V Hodgson, Jennifer Hume, Andrew Jackson, Ziad Mohid Khan, Christie Kovar-Smith, Lora R Lewis, Ryan J Lozado, Michael L Metzker, Aleksandar Milosavljevic, George R Miner, Margaret B Morgan, Lynne V Nazareth, Graham Scott, Erica Sodergren, Xing-Zhi Song, David Steffen, Sharon Wei, David A Wheeler, Mathew W Wright, Kim C Worley, Ye Yuan, Zhengdong Zhang, Charles Q Adams, M Ali Ansari-Lari, Mulu Ayele, Mary J Brown, Guan Chen, Zhijian Chen, James Clendenning, Kerstin P Clerc-Blankenburg, Runsheng Chen, Zhu Chen, Clay Davis, Oliver Delgado, Huyen H Dinh, Wei Dong, Heather Draper, Stephen Ernst, Gang Fu, Manuel L Gonzalez-Garay, Dawn K Garcia, Will Gillett, Jun Gu, Bailin Hao, Eric Haugen, Paul Havlak, Xin He, Steffen Hennig, Songnian Hu, Wei Huang, Laronda R Jackson, Leni S Jacob, Susan H Kelly, Michael Kube, Ruth Levy, Zhangwan Li, Bin Liu, Jing Liu, Wen Liu, Jing Lu, Manjula Maheshwari, Bao-Viet Nguyen, Geoffrey O Okwuonu, Anthony Palmeiri, Shiran Pasternak, Lesette M Perez, Karen A Phelps, Farah J H Plopper, Boqin Qiang, Christopher Raymond, Ruben Rodriguez, Channakhone Saenphimmachak, Jireh Santibanez, Hua Shen, Yan Shen, Sandhya Subramanian, Paul E Tabor, Daniel Verduzco, Lenee Waldron, Jian Wang, Jun Wang, Qiaoyan Wang, Gabrielle A Williams, Gane K-S Wong, Zhijian Yao, JingKun Zhang, Xiuqing Zhang, Guoping Zhao, Jianling Zhou, Yang Zhou, David Nelson, Hans Lehrach, Richard Reinhardt, Susan L Naylor, Huanming Yang, Maynard Olson, George Weinstock, Richard A Gibbs
ABSTRACT
After the completion of a draft human genome sequence, the International Human Genome Sequencing Consortium has proceeded to finish and annotate each of the 24 chromosomes comprising the human genome. Here we describe the sequencing and analysis of human chromosome 3, one of the largest human chromosomes. Chromosome 3 comprises just four contigs, one of which currently represents the longest unbroken stretch of finished DNA sequence known so far. The chromosome is remarkable in having the lowest rate of segmental duplication in the genome. It also includes a chemokine receptor gene cluster as well as numerous loci involved in multiple human cancers such as the gene encoding FHIT, which contains the most common constitutive fragile site in the genome, FRA3B. Using genomic sequence from chimpanzee and rhesus macaque, we were able to characterize the breakpoints defining a large pericentric inversion that occurred some time after the split of Homininae from Ponginae, and propose an evolutionary history of the inversion.
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
Glycerokinase from Escherichia coli, lyophilized powder, 40-100 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VI, lyophilized powder, ≥250 units/mg protein
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
β-Galactosidase from Escherichia coli, aqueous glycerol suspension, ≥500 units/mg protein (biuret)
Sigma-Aldrich
Glycerokinase from Bacillus stearothermophilus, buffered aqueous solution, ≥75 units/mg protein (biuret)
Sigma-Aldrich
Glycerokinase from Escherichia coli, 300-600 units/mL, ammonium sulfate suspension
Sigma-Aldrich
Nucleoside 5′-Diphosphate Kinase from bovine liver, buffered aqueous glycerol solution, ≥1,000 units/mg protein (biuret)
Sigma-Aldrich
β-Galactosidase from Escherichia coli, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
β-Galactosidase from bovine testes, ammonium sulfate suspension, 1.0-3.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
β-Galactosidase from bovine liver, Grade III, lyophilized powder, ≥0.15 units/mg protein
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
β-Galactosidase from Escherichia coli, suitable for enzyme immunoassay, lyophilized, powder, ~140 U/mg
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg