Merck
Past decade study of snake venom L-amino acid oxidase.
Toxicon : official journal of the International Society on Toxinology (2012-05-15)
Chunmei Guo, Shuqing Liu, Yiwen Yao, Qiaoqiao Zhang, Ming-Zhong Sun
ABSTRACT
As one of the major protein (enzyme) components of snake venom (SV), L-amino acid oxidase (LAAO) plays an important role in the toxicities and biological activities for SV. Accumulated researches in the past decade gradually revealed that SV-LAAOs induce platelet aggregation, cell apoptosis and cytotoxicity, and have anti-microbial, anti-leishmaniasis, anti-tumor and anti-HIV activity. Except for the enzymatic and structural characteristics of SV-LAAOs, the biological functions of SV-LAAOs and relevant action mechanisms are also summarized and discussed in the review. This work might provide useful inputs for future studies on SV-LAAOs.
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus atrox (Western Diamondback Rattlesnake), Type VI, dried venom
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus adamanteus, Type IV, ≥4.0 units/mg protein, aqueous suspension
Sigma-Aldrich
L-Amino Acid Oxidase from Crotalus adamanteus, Type I (dried venom)