Merck
  • Multiple concurrent collagenase clostridium histolyticum injections to Dupuytren's cords: an exploratory study.
Multiple concurrent collagenase clostridium histolyticum injections to Dupuytren's cords: an exploratory study.
BMC musculoskeletal disorders (2012-05-01)
Stephen Coleman, David Gilpin, James Tursi, Greg Kaufman, Nigel Jones, Brian Cohen
ABSTRACT
Dupuytren's contracture (DC) is a progressive fibroproliferative disorder characterized by development of nodules and collagen cords within the palmar fascia of the hand. Collagenase clostridium histolyticum (CCH) is currently approved in adults with DC for the nonsurgical treatment of a single palpable cord during a 30-day treatment cycle. This open-label pilot study was designed to examine the safety, efficacy, and multiple-dose pharmacokinetics of injecting two cords (affected joints) with multiple doses of CCH concurrently into the same hand in subjects with DC and multiple contractures. Twelve subjects with DC were enrolled, each with ≥3 contractures caused by palpable cords. Efficacy assessments were taken 30 days after treatment and adverse events (AEs) were recorded throughout. In the first treatment period, all subjects were injected with a single dose of CCH (0.58 mg) into a single cord. The same subjects entered a second treatment period 30 days later, where two different cords (affected joints) were injected concurrently on the same hand. A finger extension procedure was performed 24 hours after each administration of CCH to disrupt the enzymatically weakened cord. For metacarpophalangeal (MP) joints, mean contracture reduction per joint treated was 29.0 ± 20.7 degrees following single injection vs 30.3 ± 10.9 degrees per treated joint following multiple injections. For proximal interphalangeal (PIP) joints, mean reduction in contracture was 30.7 ± 21.1 and 22.1 ± 4.9 degrees per treated joint, respectively, for the two periods. All patients (100%) were either "quite satisfied" or "very satisfied" following either treatment cycle. The most common treatment-related AEs were edema peripheral, contusion, and pain in the treated extremity; the differences in severity for local effects of the injections were minimal between treatment periods. No serious treatment-related AEs or systemic complications were reported. These results provide preliminary evidence that two cords (affected joints) can be treated concurrently with CCH with similar efficacy and safety as cords treated individually in a sequential fashion. Multiple concurrent injections would eliminate the 30-day wait between single treatments and allow for rapid and effective treatment of patients with multiple affected joints, a significant advantage for both patient and physician. Australian New Zealand Clinical Trials Registry #ACTRN12610001045000.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type H, ≥1.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, high purity, purified by chromatography, Type VII, ≥4 FALGPA units/mg solid, lyophilized powder, ≥700 CDU/mg solid (CDU = collagen digestion units)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, powder, suitable for cell culture, ≥4 FALGPA units/mg solid, high purity, ≥700 CDU/mg solid (CDU = collagen digestion units)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, Type V-S, ≥1 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2 μm filtered solution), suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, non-sterile; 0.2 μm filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type F, ≥2.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, high purity, purified by chromatography, Type VII-S, ≥4 FALGPA units/mg solid, ≥700 CDU/mg solid (CDU = collagen digestion units)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, suitable for release of physiologically active rat hepatocytes, Type IV-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, purified by chromatography, ≥500 CDU/mg solid (CDU = collagen digestion units), lyophilized powder
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, for general use, Type I-S, 0.2-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type V, ≥1 FALGPA units/mg solid, >125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type L, ≤1.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, powder, Suitable for the digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, suitable for release of physiologically active rat epididymal adipocytes, Type II-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2μm filtered solution), 0.5-5.0 FALGPA units/mg solid, suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid