Merck
  • Livolin ameliorates elevations in alanine transaminase in HIV infected patients commencing highly active antiretroviral therapy.
Livolin ameliorates elevations in alanine transaminase in HIV infected patients commencing highly active antiretroviral therapy.
African journal of medicine and medical sciences (2013-05-16)
J A Otegbayo, M A Kuti, O Ogunbode, A E Irabor, I F Adewoles
ABSTRACT
HAART associated hepatoxicity is an important cause of poor adherence to therapy in HIV infected persons. An initial manifestation is elevation in the level ofAlanine Transaminase (ALT) in blood. We sought to evaluate the protective effects of Livolin, a phosphatidylcholine containing preparation, against elevations in this enzyme in persons just commencing HAART. All consenting patients deemed eligible for HAART and who were sero-negative for Hepatitis B and C were recruited into the study. Subjects were divided into a test group which received a thrice daily dose of Livolin capsules for 3 months in addition to HAART and a control group that received only HAART. Blood samples were collected at baseline and after 3 months and analysed for ALT, Aspartate aminotransferase, alkaline phosphatase and creatinine. The specific HAART combination, age and gender were also noted. Seventy nine (79) persons comprised of 43 test and 36 control subjects completed the study. Sixty six percent (79%) of all subjects were on Nevirapine containing combinations. In total, 8.9% and 11.7% of our patients had elevations at baseline and after 3 months respectively. These were mostly grade I, with grade II toxicity being observed in 3.3% of patients after 3 months of HAART. There was no instance of severe toxicity. For individuals with an elevation in ALT values at baseline, the mean drop at 3 months was significantly more in the test group compared with the control group (34.67 iu/L vs. 14.90 iu/L, p=0.005). Among subjects with on Nevirapine, the mean increment in ALT in the control group was 7.73 iu/L compared with 1.73 iu/L for the test group. The findings in this study mirror findings in both animal experiments and human studies of a potential benefit of phosphatidylcholine preparations, like Livolin, in protecting against drug induced hepatotoxicity.
Sigma-Aldrich
Glutamic-Oxalacetic Transaminase from porcine heart, Type I, ammonium sulfate suspension, 200-500 units/mg protein
Sigma-Aldrich
Glutamic-Pyruvic Transaminase from porcine heart, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
Glutamic-Pyruvic Transaminase from porcine heart, ammonium sulfate suspension, ≥75 units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Nevirapine
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host