Merck
  • Antioxidant properties of magnesium lithospermate B contribute to the cardioprotection against myocardial ischemia/reperfusion injury in vivo and in vitro.
Antioxidant properties of magnesium lithospermate B contribute to the cardioprotection against myocardial ischemia/reperfusion injury in vivo and in vitro.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan (2013-04-20)
Wei Quan, Ying Yin, Miaomiao Xi, Dan Zhou, Yanrong Zhu, Yue Guan, Chao Guo, Yanhua Wang, Jialin Duan, Aidong Wen
ABSTRACT
To determine the cardioprotective effect of magnesium lithospermate B (MLB) on myocardial ischemia/reperfusion (MI/R) injury and to investigate the antioxidant potential in vivo and in vitro. MI/R injury was induced by the occlusion of left anterior descending coronary artery for 30 min followed by reperfusion for 3 h in rats. After reperfusion, hearts were harvested to assess infarct size, histopathological damages, the levels of superoxide dismutase (SOD), catalase (CAT), glutathione peroxidase (GPx), reduced glutathione (GSH) and malondialdehyde (MDA). Blood samples were collected to determine serum levels of creatine kinase-MB (CK-MB), cardiac troponin (cTnl) and lactate dehydrogenase (LDH). Furthermore, simulated ischemia/reperfusion (SI/R) injury in vitro was established by oxygen and glucose deprivation (OGD) for 2 h followed by 24-hour recovery period in cardiomyocytes. The activity of LDH in the cultured supernatant and the levels of intracellular reactive oxygen species (ROS), SOD and MDA in cardiomyocytes were also measured. Finally, cardiomyocytes apoptosis was determined with flow cytometry. MLB significantly limited infarct size, ameliorated histopathological damages and prevented leakage of CK-MB, cTnI and LDH. Additionally, SOD, CAT, GPx and GSH activities were notably increased by MLB, along with the MDA content decreased as compared with the model group in rats. In vitro study, MLB also decreased LDH activity in the cultured supernatant, increased SOD activity in cardiomyocytes, reduced intracellular ROS and MDA levels, and significantly suppressed cardiomyocytes apoptosis. MLB possessed remarkably cardioprotective effects on MI/R injury in vivo and in vitro. The protection of MLB may contribute to its antioxidant properties.
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, 1000 units/mL
Sigma-Aldrich
Lactic Dehydrogenase, recombinant from E. coli, ≥90 U/mg
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine muscle, Type X, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from porcine heart, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type XVII, buffered aqueous glycerol solution, ≥400 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type III, ammonium sulfate suspension, ≥500 units/mg protein