Merck
  • Sequential selective enrichment of phosphopeptides and glycopeptides using amine-functionalized magnetic nanoparticles.
Sequential selective enrichment of phosphopeptides and glycopeptides using amine-functionalized magnetic nanoparticles.
Molecular bioSystems (2013-01-31)
Ying Zhang, Huijie Wang, Haojie Lu
ABSTRACT
As two of the most common and important post-translational modifications (PTMs) of proteins, glycosylation and phosphorylation play critical roles in biological processes. Because of the low abundance of phosphopeptides/glycopeptides, specific and sensitive strategies are especially indispensable for the identification of protein phosphorylation and glycosylation by mass spectrometry (MS). However, most of those previously reported methods only focused on enriching either phosphopeptides or glycopeptides rather than enriching both of them. In the present study, amine-functionalized magnetic nanoparticles were synthesized in a one-pot procedure and successfully used for selective enrichment of both phosphopeptides and glycopeptides. The selectivity of this method was demonstrated by analyzing the mixture of peptides/phosphopeptides/glycopeptides at molar ratio of 10:1:1; the post-enrichment recovery was 88% and 76% for phosphopeptides and glycopeptides respectively. The sensitivity was at the fmol level for both of the phosphopeptides and glycopeptides. In addition, sequence coverage was increased from 25.6% to 51.8% corresponding to a 102% increase for a model protein asialofetuin. These newly identified phosphopeptides or glycopeptides provided additional sequence information, which was beneficial to the protein identification.
Sigma-Aldrich
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid
Sigma-Aldrich
Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
Sigma-Aldrich
Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
Sigma-Aldrich
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder
Sigma-Aldrich
TrypZean® bovine, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)
Sigma-Aldrich
Trypsin from porcine pancreas, tablet, 1 mg tablet
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
Trypsin from porcine pancreas, ~1500 U/mg