Merck
  • Stable isotope labeling of N-glycosylated peptides by enzymatic deglycosylation for mass spectrometry-based glycoproteomics.
Stable isotope labeling of N-glycosylated peptides by enzymatic deglycosylation for mass spectrometry-based glycoproteomics.
Methods in molecular biology (Clifton, N.J.) (2013-01-09)
Hiroyuki Kaji, Toshiaki Isobe
ABSTRACT
Protein glycosylation is one of the most common and crucial post-translational modifications that regulates many biological processes. Because abnormal glycosylation is also associated with various pathologies, including cancer, and inflammatory and degenerative diseases, technology for comprehensive analysis of glycoproteins, or glycoproteomics, is important not only for biological studies but also for biomedical and clinical research, including the discovery of biomarkers for disease diagnosis, prognosis, and therapeutic response to drugs. Here, we describe a protocol for peptide-N-glycanase-mediated (18)O labeling of N-glycosylated peptides, termed "isotope-coded glycosylation site-specific tagging." Coupled with advanced mass spectrometry-based proteomics technology, this method facilitates the identification of hundreds to thousands of N-glycoproteins, coupled with their sites of glycosylation, from a complex biological mixture.
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, recombinant, expressed in E. coli, set of 100 units nanomolar unit
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, BioReagent, ≥95% (SDS-PAGE)
Sigma-Aldrich
Glycopeptidase A from almonds, buffered aqueous glycerol solution, ≥0.05 unit/mL
Sigma-Aldrich
PNGase F from Elizabethkingia miricola, buffered aqueous solution
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, ready-to-use solution, recombinant, expressed in E. coli