Merck
  • Oligolamellar vesicles for covalent immobilization and stabilization of D-amino acid oxidase.
Oligolamellar vesicles for covalent immobilization and stabilization of D-amino acid oxidase.
Enzyme and microbial technology (2012-12-04)
Makoto Yoshimoto, Miku Yamasaki, Masakazu Okamoto, Hiroshi Umakoshi, Ryoichi Kuboi
ABSTRACT
Oligolamellar phospholipid vesicles incorporated with d-amino acid oxidase from porcine kidney (OV-DAO) were prepared by encapsulating pre-formed enzyme-bound unilamellar vesicles (UV-DAO) with bilayers of 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine (POPC). The bilayer of UV-DAO was composed of POPC, 30 mol% of cholesterol and 15 mol% of 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(glutaryl) (NGPE) that was responsible for covalent linking to D-amino acid oxidase (DAO). OV-DAO and UV-DAO showed the activity to catalyze the oxidation of D-alanine as measured based on the hydrogen peroxide produced. The oligolamellar and unilamellar structure of OV-DAO and UV-DAO, respectively was elucidated based on the quenching characteristics of bilayers-incorporated fluorescent lipid 7-nitro-2,1,3-benzoxadiazol-4-yl-phosphoethanolamine (NBD-PE) and the size distribution of the vesicles measured with the dynamic light scattering method. The enzyme activity of OV-DAO and UV-DAO was significantly stabilized at 50°C compared to that of free DAO at the fixed enzyme concentration of 3.29 μg/mL. At the temperature, OV-DAO and UV-DAO showed the remaining activity of 52.7 and 29.6%, respectively at the incubation time of 20 min while free DAO was completely deactivated. Thus the dimeric form of DAO could be stabilized by its coupling to the surface of UV-DAO membrane being the inner bilayer of OV-DAO. Furthermore, the thermal denaturation of DAO and dissociation of flavin adenine dinucleotide (FAD) from the subunits of enzyme were prevented in the aqueous phase formed between the bilayers of OV-DAO.
Sigma-Aldrich
D-Amino Acid Oxidase from porcine kidney, powder, ≥1.5 units/mg solid
Pricing and availability is not currently available.