Merck
Effect of vanadium chloroperoxidase on Enterococcus faecalis biofilms.
Journal of endodontics (2011-12-14)
Ilona F Persoon, Michel A Hoogenkamp, Aleksandra Bury, Paul R Wesselink, Aloysius F Hartog, Ronald Wever, Wim Crielaard
ABSTRACT
The aim of this study was to explore the antimicrobial effect of vanadium chloroperoxidase (VCPO) reaction products on Enterococcus faecalis biofilms of 4 different strains. Twenty-four-hour biofilms of E. faecalis strains V583, ER5/1, E2, and OS-16 were incubated in mixtures with VCPO, halide (either bromide or chloride), and hydrogen peroxide. The antibacterial efficacy was assessed by colony-forming unit counts. The VCPO reaction products had a similar efficacy in reducing the viability of the 4 strains of E. faecalis (94%; range, 87%-100%). Bromide as the halogen of choice was more effective on E. faecalis strains E2 and OS-16, as compared with chloride (Mann-Whitney U test; P < .05). Despite different quantities of produced biofilms by the 4 strains, VCPO treatment was similarly effective toward all strains (Kruskal-Wallis test; P < .05). VCPO treatment results in an antimicrobial effect toward in vitro E. faecalis biofilms and might provide an addition to current endodontic treatment, possibly as an antimicrobial dressing.
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, aqueous suspension, brown, >10,000 U/mL
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, buffered aqueous suspension, 1,000-2,000 units/mg protein (E1%/280)
Sigma-Aldrich
Chloroperoxidase from Caldariomyces fumago, buffered aqueous suspension, ≥3,000 units/mL