Merck
  • Construction, expression and characterization of fusion enzyme from Arthrobacter oxydans dextranase and Klebsiella pneumoniae amylase.
Construction, expression and characterization of fusion enzyme from Arthrobacter oxydans dextranase and Klebsiella pneumoniae amylase.
Biotechnology letters (2009-03-24)
Young-Min Kim, Eun-Ah Ko, Hee-Kyoung Kang, Doman Kim
ABSTRACT
An artificial fusion protein of Arthrobacter oxydans dextranase and Klebsiella pneumoniae alpha-amylase was constructed and expressed in Escherichia coli. Most of the expressed protein existed as an insoluble fraction, which was solubilized with urea. The purified fusion enzyme electrophoretically migrated as a single protein band; M = 137 kDa, and exhibited activities of both dextranase (10.8 U mg(-1)) and amylase (7.1 U mg(-1)), which were lower than that of reference dextranase (13.3 U mg(-1)) and alpha-amylase (103 U mg(-1)). The fusion enzyme displayed bifunctional enzyme activity at pH 5-7 at 37 degrees C. These attributes potentially make the fusion enzyme more convenient for use in sugar processing than a two-enzyme system.
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 100-250 units/mg protein
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 10-25 units/mg solid
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 400-800 units/mg protein
Sigma-Aldrich
Dextranase from Chaetomium erraticum