Merck
  • E2F1, ARID3A/Bright and Oct-2 factors bind to the Epstein-Barr virus C promoter, EBNA1 and oriP, participating in long-distance promoter-enhancer interactions.
E2F1, ARID3A/Bright and Oct-2 factors bind to the Epstein-Barr virus C promoter, EBNA1 and oriP, participating in long-distance promoter-enhancer interactions.
The Journal of general virology (2012-02-04)
Cecilia Boreström, Alma Forsman, Ulla Rüetschi, Lars Rymo
ABSTRACT
The Epstein-Barr virus (EBV) C promoter (Cp) regulates several genes required for B-cell proliferation in latent EBV infection. The family of repeats (FR) region of the latent origin of plasmid replication (oriP) functions as an Epstein-Barr nuclear antigen 1 (EBNA1)-dependent distant enhancer of Cp activity, and the enhancer-promoter interaction is mediated by a higher-order multi-protein complex containing several copies of EBNA1. Using DNA-affinity purification with a 170 bp region of the Cp in combination with mass spectrometry, we identified the cell cycle-regulatory protein E2F1, the E2F-binding protein ARID3A, and the B-cell-specific transcription factor Oct-2 as components of this multi-protein complex. Binding of the three factors to the FR region of oriP was determined by DNA-affinity and immunoblot analysis. Co-immunoprecipitation and proximity ligation analysis revealed that the three factors, E2F1, ARID3A and Oct-2, interact with each other as well as with EBNA1 in the nuclei of EBV-positive cells. Using the chromatin immunoprecipitation assay, we showed that E2F1 and Oct-2 interacted with the FR part of oriP and the Cp, but the ARID3A interaction was, however, only detected at the Cp. Our findings support the hypothesis that EBNA1 initiates transcription at the Cp via interactions between multiple EBNA1 homodimers and cellular transcription factors in a large molecular machinery that forms a dynamic interaction between Cp and FR.
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)