Merck
  • Antiproliferative and antitumor activities of D-reverse peptides derived from the second type-1 repeat of thrombospondin-1.
Antiproliferative and antitumor activities of D-reverse peptides derived from the second type-1 repeat of thrombospondin-1.
The journal of peptide research : official journal of the American Peptide Society (1997-10-06)
N H Guo, H C Krutzsch, J K Inman, C S Shannon, D D Roberts
ABSTRACT
The extracellular matrix glycoprotein thrombospondin-1 (TSP1) inhibits angiogenesis, endothelial cell growth, motility and adhesion. Peptides from the type I repeats of TSP1 mimic the adhesive and growth inhibitory activities of the intact protein and specifically interact with heparin and transforming growth factor-beta (TGF beta). To define the structural basis for the antiangiogenic activities of these peptides, we prepared analogs of the TSP1 peptide KRFKQDGGWSHWSPWSSC. L-forward, L-reverse, and D-reverse (retro-inverso) analogs displayed identical activities for binding to heparin, demonstrating a lack of stereospecificity for heparin binding. The L-reverse and D-reverse peptides, however, had somewhat decreased abilities to activate latent TGF beta. Conjugation of the forward peptides through a C-terminal thioether and the reverse peptides through an N-terminal thioether to polysucrose abolished the adhesive activity of the peptides and enhanced their antiproliferative activities for endothelial and breast carcinoma cells stimulated by fibroblast growth factor-2. Their antiproliferative activities were independent of latent TGF beta activation, because substitution of an Ala residue for the essential Phe residue in the TSP1 type-1 repeat peptide increased their potency for inhibiting TSP1 binding to heparin and for inhibiting endothelial cell proliferation. Although the conjugated peptides were inactive in vivo, an unconjugated retro-inverso analog of the native TSP peptide inhibited breast tumor growth in a mouse xenograft model. Thus, these TSP-derived peptide analogs antagonize endothelial growth through their heparin-binding activity rather than through activation of latent TGF beta or increasing cell adhesion. These stable analogs may therefore be useful as therapeutic inhibitors of angiogenesis stimulated by fibroblast growth factor-2.
Sigma-Aldrich
Polysucrose 400, powder
Pricing and availability is not currently available.