Merck
  • Human high temperature requirement serine protease A1 (HTRA1) degrades tau protein aggregates.
Human high temperature requirement serine protease A1 (HTRA1) degrades tau protein aggregates.
The Journal of biological chemistry (2012-04-27)
Annette Tennstaedt, Simon Pöpsel, Linda Truebestein, Patrick Hauske, Anke Brockmann, Nina Schmidt, Inga Irle, Barbara Sacca, Christof M Niemeyer, Roland Brandt, Hanna Ksiezak-Reding, Anca Laura Tirniceriu, Rupert Egensperger, Alfonso Baldi, Leif Dehmelt, Markus Kaiser, Robert Huber, Tim Clausen, Michael Ehrmann
ABSTRACT
Protective proteases are key elements of protein quality control pathways that are up-regulated, for example, under various protein folding stresses. These proteases are employed to prevent the accumulation and aggregation of misfolded proteins that can impose severe damage to cells. The high temperature requirement A (HtrA) family of serine proteases has evolved to perform important aspects of ATP-independent protein quality control. So far, however, no HtrA protease is known that degrades protein aggregates. We show here that human HTRA1 degrades aggregated and fibrillar tau, a protein that is critically involved in various neurological disorders. Neuronal cells and patient brains accumulate less tau, neurofibrillary tangles, and neuritic plaques, respectively, when HTRA1 is expressed at elevated levels. Furthermore, HTRA1 mRNA and HTRA1 activity are up-regulated in response to elevated tau concentrations. These data suggest that HTRA1 is performing regulated proteolysis during protein quality control, the implications of which are discussed.
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg
Sigma-Aldrich
N-Succinyl-Leu-Leu-Val-Tyr-7-Amido-4-Methylcoumarin, ≥90% (HPLC)
Sigma-Aldrich
Tetrahydrothiophene, 99%