Merck
  • Structural basis for VLDLR recognition by eastern equine encephalitis virus.
Structural basis for VLDLR recognition by eastern equine encephalitis virus.
Nature communications (2024-08-03)
Pan Yang, Wanyu Li, Xiaoyi Fan, Junhua Pan, Colin J Mann, Haley Varnum, Lars E Clark, Sarah A Clark, Adrian Coscia, Himanish Basu, Katherine Nabel Smith, Vesna Brusic, Jonathan Abraham
ABSTRACT
Eastern equine encephalitis virus (EEEV) is the most virulent alphavirus that infects humans, and many survivors develop neurological sequelae, including paralysis and intellectual disability. Alphavirus spike proteins comprise trimers of heterodimers of glycoproteins E2 and E1 that mediate binding to cellular receptors and fusion of virus and host cell membranes during entry. We recently identified very-low density lipoprotein receptor (VLDLR) and apolipoprotein E receptor 2 (ApoER2) as cellular receptors for EEEV and a distantly related alphavirus, Semliki Forest virus (SFV). Here, we use single-particle cryo-electron microscopy (cryo-EM) to determine structures of the EEEV and SFV spike glycoproteins bound to the VLDLR ligand-binding domain and found that EEEV and SFV interact with the same cellular receptor through divergent binding modes. Our studies suggest that the ability of LDLR-related proteins to interact with viral spike proteins through very small footprints with flexible binding modes results in a low evolutionary barrier to the acquisition of LDLR-related proteins as cellular receptors for diverse sets of viruses.
Sigma-Aldrich
Anti-Eastern Equine Encephalitis Antibody, Virus, clone 1A4B.6, clone 1A4B.6, Chemicon®, from mouse